N,N-Diethylhydroxylamine ∼85% ∼8.6 mol/L in water

Supplier: TCI America

D1178-500ML D1178-25ML
TCD1178-500MLEA 94.4 USD
TCD1178-500ML TCD1178-25ML
N,N-Diethylhydroxylamine ∼85% ∼8.6 mol/L in water
N,N-Diethylhydroxylamine

CAS Number: 3710-84-7
MDL Number: MFCD00002126
Molecular Formula: C4H11NO
Molecular Weight: 89.14
Form: Clear Liquid
Color: Colorless
Flash Point (°C): 45
Specific Gravity (20/20): 0.90

Formula: C₄H₁₁NO
Density: 0.90 (20 °C)
Flash Pt: 45 °C
MDL Number: MFCD00002126
CAS Number: 3710-84-7
EINECS: 223-055-4
https://us.vwr.com/stibo/web/std.lang.all/81/01/28978101.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR