ω,ω-Dichloroacetophenone ≥97.0%

Supplier: TCI America

Synonyms: 2,2-Dichloroacetophenone α,α-Dichloroacetophenone

D1122-25G
TCD1122-025GEA 210.01 USD
TCD1122-025G
ω,ω-Dichloroacetophenone ≥97.0%
ω,ω-Dichloroacetophenone

CAS Number: 2648-61-5
MDL Number: MFCD00000844
Molecular Formula: C8H6Cl2O
Molecular Weight: 189.04
Purity/Analysis Method: 97.0% (GC)
Form: Clear Liquid
Boiling point (°C): 247
Flash Point (°C): 110
Freezing point (°C): 18
Specific Gravity (20/20): 1.34

Formula: (C₆H₅)C(O)CHCl₂
MW: 189.04 g/mol
Boiling Pt: 247 °C
Melting Pt: 23…24 °C
Density: 1.34 (20 °C)
Flash Pt: 110 °C
MDL Number: MFCD00000844
CAS Number: 2648-61-5
https://us.vwr.com/stibo/web/std.lang.all/63/56/28976356.jpg

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR