1H,1H,7H-Dodecafluoro-1-heptanol ≥97.0%

Supplier: TCI America

Synonyms: 1H,1H,7H-Perfluoro-1-heptanol2,2,3,3,4,4,5,5,6,6,7,7-Dodecafluoro-1-heptanol1H,1h,7h-dodecafluoro-1-heptanol

D1101-25G
TCD1101-25GEA 44.66 USD
TCD1101-25G
1H,1H,7H-Dodecafluoro-1-heptanol ≥97.0%
1H,1H,7H-Dodecafluoro-1-heptanol

CAS Number: 335-99-9
MDL Number: MFCD00039630
Molecular Formula: C7H4F12O
Molecular Weight: 332.09
Purity/Analysis Method: 97.0% (GC)
Form: Clear Liquid
Boiling point (°C): 170
Melting point (°C): -20
Specific Gravity (20/20): 1.75


Product Categories: Synthetic Organic Chemistry, Fluorous Compounds, Fluorinated Building Blocks
Other Product Names / Synonyms: 1,1,7-Trihydroperfluoroheptanol

Formula: C₇H₄F₁₂O
MW: 332.09 g/mol
Boiling Pt: 170 °C
Melting Pt: –20 °C
Density: 1.75 (20 °C)
MDL Number: MFCD00039630
CAS Number: 335-99-9
EINECS: 206-406-6
https://us.vwr.com/stibo/web/std.lang.all/63/38/28976338.jpg

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR