4,7-Dichloroquinoline ≥98.0%

Supplier: TCI America
Warning

D1100-25G
TCD1100-025GEA 42 USD
TCD1100-025G
4,7-Dichloroquinoline ≥98.0%
4,7-Dichloroquinoline

CAS Number: 86-98-6
MDL Number: MFCD00006774
Molecular Formula: C9H5Cl2N
Molecular Weight: 198.05
Purity/Analysis Method: 98.0% (GC)
Form: Crystal
Boiling point (°C): 148
Melting point (°C): 87
Flash Point (°C): 164

Formula: C₉H₅Cl₂N
MW: 198.05 g/mol
Boiling Pt: 148 °C
Melting Pt: 87 °C
Flash Pt: 164 °C
Storage Temperature: Ambient
MDL Number: MFCD00006774
CAS Number: 86-98-6
EINECS: 201-714-7
https://us.vwr.com/stibo/web/std.lang.all/63/37/28976337.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR