4,4'-Bis(hexyloxy)azoxybenzene ≥96.0% (by total nitrogen basis)
Supplier: TCI America
Synonyms:
4,4'-Dihexyloxyazoxybenzene
D1092-1G
TCD1092-001GEA
649
USD
TCD1092-001G
4,4'-Bis(hexyloxy)azoxybenzene ≥96.0% (by total nitrogen basis)
4,4'-Bis(hexyloxy)azoxybenzene
CAS Number: 2587-42-0
MDL Number: MFCD00043672
Molecular Formula: C24H34N2O3
Molecular Weight: 398.55
Purity/Analysis Method:
Form: Crystal
Product Categories: Materials Chemistry, Liquid Crystal Materials (Optically Functional Materials)
Other Product Names / Synonyms: 4,4'-Dihexyloxyazoxybenzene
Formula:
C₂₄H₃₄N₂O₃ MW: 398.55 g/mol |
MDL Number:
MFCD00043672 CAS Number: 2587-42-0 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...