3,5-Dibromosalicylic acid ≥98.0% (by titrimetric analysis)

Supplier: TCI America
Warning

Synonyms: 3,5-Dibromo-2-hydroxybenzoic acid

D1087-25G D1087-500G
TCD1087-025GEA 48.01 USD
TCD1087-025G TCD1087-500G
3,5-Dibromosalicylic acid ≥98.0% (by titrimetric analysis)
3,5-Dibromosalicylic acid

CAS Number: 3147-55-5
MDL Number: MFCD00002441
Molecular Formula: C7H4Br2O3
Molecular Weight: 295.91
Purity/Analysis Method: 98.0% (T)
Form: Crystal
Melting point (°C): 223

Formula: C₇H₄Br₂O₃
MW: 295.91 g/mol
Boiling Pt: 365 °C (1013 hPa)
Melting Pt: 223 °C
Storage Temperature: Ambient
MDL Number: MFCD00002441
CAS Number: 3147-55-5
EINECS: 221-570-9
Merck Index: 13,03053
https://us.vwr.com/stibo/web/std.lang.all/63/27/28976327.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR