Diphenyl chlorophosphate ≥95.0%
Supplier: TCI America
Synonyms:
Diphenyl phosphorochloridate
D1059-500G
D1059-25G
D1059-100G
TCD1059-25GEA
21.64
USD
TCD1059-25G
TCD1059-100G
TCD1059-500G
Diphenyl chlorophosphate ≥95.0%
Diphenyl chlorophosphate
CAS Number: 2524-64-3
MDL Number: MFCD00003030
Molecular Formula: C12H10ClO3P
Molecular Weight: 268.63
Purity/Analysis Method:
Form: Clear Liquid
Boiling point (°C): 316
Flash Point (°C): 113
Specific Gravity (20/20): 1.30
Product Categories: Synthetic Organic Chemistry, Phosphorylation, Nucleic Acid Synthesis
Other Product Names / Synonyms: Chlorodiphenyl Phosphate, Diphenyl Phosphorochloridate, Diphenylphosphoryl Chloride, Phosphorochloridic Acid Diphenyl Ester
Formula:
C₁₂H₁₀ClO₃P Boiling Pt: 316 °C Density: 1.30 (20 °C) Flash Pt: 113 °C |
MDL Number:
MFCD00003030 CAS Number: 2524-64-3 EINECS: 219-759-6 UN: 3265 ADR: 8,III |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...