1,2-Diiodoethane ≥97.0%

Supplier: TCI America
Warning

D1025-25G
TCD1025-025GEA 58.54 USD
TCD1025-025G
1,2-Diiodoethane ≥97.0%
1,2-Diiodoethane

CAS Number: 624-73-7
MDL Number: MFCD00001093
Molecular Formula: C2H4I2
Molecular Weight: 281.86
Purity/Analysis Method: 97.0% (GC)
Form: Crystal
Melting point (°C): 82
Storage Temperature: 0°C


Other Product Names / Synonyms: Ethylene Diiodide, Ethylene Iodide

Formula: C₂H₄I₂
MW: 281.86 g/mol
Boiling Pt: 206 °C (1013 hPa)
Melting Pt: 82 °C
Density: 2.132 g/cm³ (20 °C)
Storage Temperature: Refrigerator
MDL Number: MFCD00001093
CAS Number: 624-73-7
EINECS: 210-859-5
https://us.vwr.com/stibo/web/std.lang.all/62/30/28976230.jpg

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR