2,5-Dimethoxytetrahydrofuran ≥98.0%

Supplier: TCI America

D0986-500G D0986-25G D0986-100G
TCD0986-100GEA 97.44 USD
TCD0986-100G TCD0986-500G TCD0986-25G
2,5-Dimethoxytetrahydrofuran ≥98.0%
2,5-Dimethoxytetrahydrofuran

CAS Number: 696-59-3
MDL Number: MFCD00005359
Molecular Formula: C6H12O3
Molecular Weight: 132.16
Purity/Analysis Method: 98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 146
Flash Point (°C): 35
Specific Gravity (20/20): 1.03


Other Product Names / Synonyms: Tetrahydro-2,5-dimethoxyfuran

Formula: C₆H₁₂O₃
MW: 132.16 g/mol
Boiling Pt: 146 °C
Melting Pt: –45 °C
Density: 1.03 (20 °C)
Flash Pt: 35 °C
Storage Temperature: Ambient
MDL Number: MFCD00005359
CAS Number: 696-59-3
EINECS: 211-797-1
UN: 3271
ADR: 3,III
https://us.vwr.com/stibo/web/std.lang.all/61/68/28976168.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR