Dipropyl sulfite ≥99.0%
Supplier: TCI America
|
Warning
|
D0935-25ML
TCD0935-025MLEA
211.06
USD
TCD0935-025ML
Dipropyl sulfite ≥99.0%
Dipropyl sulphite
CAS Number: 623-98-3
MDL Number: MFCD00059404
Molecular Formula: C6H14O3S
Molecular Weight: 166.24
Purity/Analysis Method:
Form: Clear Liquid
Boiling point (°C): 192
Flash Point (°C): 110
Specific Gravity (20/20): 1.03
Formula:
C₆H₁₄O₃S MW: 166.24 g/mol Boiling Pt: 192 °C Density: 1.03 (20 °C) Flash Pt: 110 °C |
MDL Number:
MFCD00059404 CAS Number: 623-98-3 UN: 3334 ADR: 9, |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...