5,5-Diphenylhydantoin ≥99.0% (by titrimetric analysis)

Supplier: TCI America

Synonyms: Phenytoin

D0894-25G
TCD0894-025GEA 58.09 USD
TCD0894-025G
5,5-Diphenylhydantoin ≥99.0% (by titrimetric analysis)
5,5-Diphenylhydantoin

CAS Number: 57-41-0
MDL Number: MFCD00005264
Molecular Formula: C15H12N2O2
Molecular Weight: 252.27
Purity/Analysis Method: 99.0% (T)
Form: Crystal
Melting point (°C): 298


Product Categories: Bioscience, Neurotransmitter Agent Ingredients
Other Product Names / Synonyms: Phenytoin

Formula: C₁₅H₁₂N₂O₂
Melting Pt: 298 °C
MDL Number: MFCD00005264
CAS Number: 57-41-0
EINECS: 200-328-6
https://us.vwr.com/stibo/web/std.lang.all/60/13/28976013.jpg

Order Now

Specification Test Results

Purity/Analysis Method 99.0% (T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR