1,3-Diphenylguanidine ≥98.0% (by titrimetric analysis)
Supplier: TCI America
|
Warning
|
Synonyms:
N,N'-Diphenylguanidine
, sym-Diphenylguanidine
D0892-500G
D0892-25G
TCD0892-25GEA
20.3
USD
TCD0892-25G
TCD0892-500G
1,3-Diphenylguanidine ≥98.0% (by titrimetric analysis)
1,3-Diphenylguanidine
CAS Number: 102-06-7
MDL Number: MFCD00001758
Molecular Formula: C13H13N3
Molecular Weight: 211.27
Purity/Analysis Method:
Form: Crystal
Melting point (°C): 147
Flash Point (°C): 170
Formula:
C₁₃H₁₃N₃ MW: 211.27 g/mol Boiling Pt: 321 °C (1013 hPa) Melting Pt: 147 °C Density: 1.19 g/cm³ (20 °C) Flash Pt: 170 °C Storage Temperature: Ambient |
MDL Number:
MFCD00001758 CAS Number: 102-06-7 EINECS: 203-002-1 UN: 2811 ADR: 6.1,III Merck Index: 13,03356 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...