Sodium 2,4-dinitrobenzenesulfonate ≥98.0% (by HPLC, titration analysis)

Supplier: TCI America

Synonyms: 2,4-Dinitrobenzenesulphonic acid sodium salt

D0821-500G D0821-25G
TCD0821-25GEA 68.01 USD
TCD0821-25G TCD0821-500G
Sodium 2,4-dinitrobenzenesulfonate ≥98.0% (by HPLC, titration analysis)
Sodium 2,4-dinitrobenzenesulphonate

CAS Number: 885-62-1
MDL Number: MFCD00007471
Molecular Formula: C6H4N2O7S
Molecular Weight: 270.15
Purity/Analysis Method: 98.0% (HPLC,T)
Form: Crystal


Other Product Names / Synonyms: 2,4-Dinitrobenzenesulfonic Acid Sodium Salt

Formula: C₆H₃N₂NaO₇S
MW: 270.15 g/mol
MDL Number: MFCD00007471
CAS Number: 885-62-1
EINECS: 212-943-7
https://us.vwr.com/stibo/web/std.lang.all/59/10/28975910.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR