1,1-Dimethylurea ≥98.0% (by total nitrogen basis)
Supplier: TCI America
Synonyms:
N,N-Dimethylurea
D0809-500G
D0809-25G
TCD0809-025GEA
84.25
USD
TCD0809-025G
TCD0809-500G
1,1-Dimethylurea ≥98.0% (by total nitrogen basis)
1,1-Dimethylurea
CAS Number: 598-94-7
MDL Number: MFCD00007959
Molecular Formula: C3H8N2O
Molecular Weight: 88.11
Purity/Analysis Method:
Form: Crystal
Melting point (°C): 182
Formula:
C₃H₈N₂O MW: 88.11 g/mol Melting Pt: 182 °C |
MDL Number:
MFCD00007959 CAS Number: 598-94-7 EINECS: 209-957-0 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...