1,3-Diphenylguanidine ≥98.0% (by HPLC, titration analysis)

Supplier: TCI America
Warning

Synonyms: N,N'-Diphenylguanidinesym-Diphenylguanidine

D0723-25G D0723-500G
TCD0723-25GEA 22.78 USD
TCD0723-25G TCD0723-500G
1,3-Diphenylguanidine ≥98.0% (by HPLC, titration analysis)
1,3-Diphenylguanidine

CAS Number: 102-06-7
MDL Number: MFCD00001758
Molecular Formula: C13H13N3
Molecular Weight: 211.27
Purity/Analysis Method: 98.0% (HPLC,T)
Form: Crystal
Melting point (°C): 147
Flash Point (°C): 159

Formula: C₁₃H₁₃N₃
MW: 211.27 g/mol
Boiling Pt: 321 °C (1013 hPa)
Melting Pt: 147 °C
Density: 1.19 g/cm³ (20 °C)
Flash Pt: 159 °C
Storage Temperature: Ambient
MDL Number: MFCD00001758
CAS Number: 102-06-7
EINECS: 203-002-1
UN: 2811
ADR: 6.1,III
Merck Index: 13,03356
https://us.vwr.com/stibo/web/std.lang.all/56/91/28975691.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR