3,4-Xylidine ≥99.0% (by GC, titration analysis)

Supplier: TCI America
Danger

Synonyms: 3,4-Dimethylaniline4-Amino-o-xylene

D0670-500G D0670-25G
TCD0670-500GEA 269 USD
TCD0670-500G TCD0670-25G
3,4-Xylidine ≥99.0% (by GC, titration analysis)
3,4-Xylidine

CAS Number: 95-64-7
MDL Number: MFCD00007810
Molecular Formula: C8H11N
Molecular Weight: 121.18
Purity/Analysis Method: 99.0% (GC,T)
Form: Crystal
Boiling point (°C): 226
Melting point (°C): 51
Flash Point (°C): 107


Other Product Names / Synonyms: 3,4-Xylidine

Formula: C₈H₁₁N
MW: 121.18 g/mol
Boiling Pt: 226 °C
Melting Pt: 51 °C
Density: 1.0755 g/cm³ (18 °C)
Flash Pt: 107 °C
Storage Temperature: Ambient
MDL Number: MFCD00007810
CAS Number: 95-64-7
EINECS: 202-437-4
UN: 3452
ADR: 6.1,II
Merck Index: 12,10217
https://us.vwr.com/stibo/web/std.lang.all/55/65/28975565.jpg

Order Now

Specification Test Results

Purity/Analysis Method 99.0% (GC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR