2,5-Xylidine ≥97.0%

Supplier: TCI America
Danger

Synonyms: 2,5-Dimethylaniline2-Amino-p-xylene

D0668-25G D0668-500G
TCD0668-500GEA 228 USD
TCD0668-500G TCD0668-25G
2,5-Xylidine ≥97.0%
2,5-Xylidine

CAS Number: 95-78-3
MDL Number: MFCD00007743
Molecular Formula: C8H11N
Molecular Weight: 121.18
Purity/Analysis Method: 97.0% (GC)
Form: Clear Liquid
Boiling point (°C): 104
Flash Point (°C): 106
Freezing point (°C): 11
Specific Gravity (20/20): 0.98


Other Product Names / Synonyms: 2,5-Xylidine, p-Xylidine

Formula: C₈H₁₁N
MW: 121.18 g/mol
Boiling Pt: 104 °C
Melting Pt: 15 °C
Density: 0.98 (20 °C)
Flash Pt: 106 °C
Storage Temperature: Ambient
MDL Number: MFCD00007743
CAS Number: 95-78-3
EINECS: 202-451-0
UN: 1711
ADR: 6.1,II
Merck Index: 12,10217
https://us.vwr.com/stibo/web/std.lang.all/55/59/28975559.jpg

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR