2,4-Xylidine ≥98.0% (by GC, titration analysis)

Supplier: TCI America
Danger

Synonyms: 2,4-Dimethylanilinem-Xylidine2,4-Xylidine4-Amino-m-xylene

D0667-25ML D0667-500ML
TCD0667-500MLEA 215.18 USD
TCD0667-500ML TCD0667-25ML
2,4-Xylidine ≥98.0% (by GC, titration analysis)
2,4-Xylidine

CAS Number: 95-68-1
MDL Number: MFCD00007738
Molecular Formula: C8H11N
Molecular Weight: 121.18
Purity/Analysis Method: 98.0% (GC,T)
Form: Clear Liquid
Boiling point (°C): 214
Melting point (°C): 16
Flash Point (°C): 90
Specific Gravity (20/20): 0.98


Other Product Names / Synonyms: 2,4-Xylidine, m-Xylidine

Formula: C₈H₁₁N
MW: 121.18 g/mol
Boiling Pt: 214 °C
Melting Pt: 16 °C
Density: 0.98 (20 °C)
Flash Pt: 90 °C
Storage Temperature: Ambient
MDL Number: MFCD00007738
CAS Number: 95-68-1
EINECS: 202-440-0
UN: 1711
ADR: 6.1,II
Merck Index: 12,10217
https://us.vwr.com/stibo/web/std.lang.all/55/57/28975557.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR