2,3-Xylidine ≥99.0% (by GC, titration analysis)

Supplier: TCI America
Danger

Synonyms: 3-Amino-o-xylene2,3-Xylidine

D0666-25G D0666-500G
TCD0666-500GEA 96 USD
TCD0666-500G TCD0666-025G
2,3-Xylidine ≥99.0% (by GC, titration analysis)
2,3-Dimethylaniline

CAS Number: 87-59-2
MDL Number: MFCD00007732
Molecular Formula: C8H11N
Molecular Weight: 121.18
Purity/Analysis Method: 99.0% (GC,T)
Form: Clear Liquid
Boiling point (°C): 114
Flash Point (°C): 97
Freezing point (°C): 3
Specific Gravity (20/20): 1.00


Other Product Names / Synonyms: 2,3-Xylidine, vic-o-Xylidine

Formula: C₈H₁₁N
MW: 121.18 g/mol
Boiling Pt: 114 °C
Melting Pt: –15 °C
Density: 1.00 (20 °C)
Flash Pt: 97 °C
Storage Temperature: Ambient
MDL Number: MFCD00007732
CAS Number: 87-59-2
EINECS: 201-755-0
UN: 1711
ADR: 6.1,II
Merck Index: 12,10217
https://us.vwr.com/stibo/web/std.lang.all/55/55/28975555.jpg

Order Now

Specification Test Results

Purity/Analysis Method 99.0% (GC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR