2,4-Dimethoxybenzaldehyde ≥98.0%
Supplier: TCI America
D0626-100G
D0626-25G
D0626-500G
TCD0626-025GEA
151.91
USD
TCD0626-025G
TCD0626-500G
TCD0626-100G
2,4-Dimethoxybenzaldehyde ≥98.0%
2,4-Dimethoxybenzaldehyde
CAS Number: 613-45-6
MDL Number: MFCD00003311
Molecular Formula: C9H10O3
Molecular Weight: 166.18
Purity/Analysis Method:
Form: Crystal
Boiling point (°C): 165
Melting point (°C): 71
Formula:
C₉H₁₀O₃ MW: 166.18 g/mol Boiling Pt: 165 °C Melting Pt: 71 °C Storage Temperature: Ambient |
MDL Number:
MFCD00003311 CAS Number: 613-45-6 EINECS: 210-342-4 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...