1,5-Dihydroxy-9,10-anthraquinone ≥85.0% (by HPLC)
Supplier: TCI America
|
Warning
|
Synonyms:
1,5-Dihydroxyanthraquinone
, Anthrarufin
D0562-500G
D0562-25G
TCD0562-500GEA
160.94
USD
TCD0562-500G
TCD0562-025G
1,5-Dihydroxy-9,10-anthraquinone ≥85.0% (by HPLC)
1,5-Dihydroxy-9,10-anthraquinone
CAS Number: 117-12-4
MDL Number: MFCD00001210
Molecular Formula: C14H8O4
Molecular Weight: 240.21
Purity/Analysis Method:
Form: Crystal
Melting point (°C): 280
Other Product Names / Synonyms: 1,5-Dihydroxyanthraquinone
Formula:
C₁₄H₈O₄ MW: 240.22 g/mol Melting Pt: 280 °C |
MDL Number:
MFCD00001210 CAS Number: 117-12-4 EINECS: 204-175-6 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...