TCIAD0467

D0467-25G D0467-500G
TCD0467-500GEA 158.07 USD
TCD0467-500G TCD0467-025G
TCIAD0467
CAS Number: 14426-20-1
MDL Number: MFCD00036374
Molecular Formula: C6H15NO
Molecular Weight: 153.65
Purity/Analysis Method: 98.0% (T)
Form: Crystal
Melting point (°C): 133

Other Product Names / Synonyms: N,N-Diethylethanolamine Hydrochloride
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR