1,3-Diethoxy-2-propanol ≥98.0%

Supplier: TCI America

D0460-10ML
TCD0460-010MLEA 96.39 USD
TCD0460-010ML
1,3-Diethoxy-2-propanol ≥98.0%
1,3-Diethoxy-2-propanol

CAS Number: 4043-59-8
MDL Number: MFCD00026934
Molecular Formula: C7H16O3
Molecular Weight: 148.20
Purity/Analysis Method: 98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 191
Flash Point (°C): 58
Specific Gravity (20/20): 0.95


Other Product Names / Synonyms: Glycerol alpha,alpha'-Diethyl Ether

Formula: C₇H₁₆O₃
MW: 148.2 g/mol
Boiling Pt: 191 °C
Density: 0.95 (20 °C)
Flash Pt: 58 °C
MDL Number: MFCD00026934
CAS Number: 4043-59-8
EINECS: 223-736-6

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR