Dicyclohexylcarbodiimide 25% ∼1.2 mol/L in pyridine
Supplier: TCI America
|
Danger
|
Synonyms:
N,N'-Dicyclohexylcarbodiimide
D0437-100ML
TCD0437-100MLEA
185.75
USD
TCD0437-100ML
Dicyclohexylcarbodiimide 25% ∼1.2 mol/L in pyridine
Dicyclohexylcarbodiimide
CAS Number: 538-75-0
MDL Number: MFCD00011659
Molecular Formula: C13H22N2
Molecular Weight: 206.33
Form: Clear Liquid
Color: Colorless
Specific Gravity (20/20): 0.98
Storage Temperature: 0-10°C
Other Product Names / Synonyms: DCC
Formula:
C₁₃H₂₂N₂ MW: 206.33 g/mol Boiling Pt: 148…152 °C (15 hPa) Melting Pt: 35…36 °C Density: 0.98 (20 °C) Flash Pt: 113 °C |
MDL Number:
MFCD00011659 CAS Number: 538-75-0 EINECS: 208-704-1 UN: 2811 ADR: 6.1,III Merck Index: 13,03123 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...