α,2-Dichlorotoluene ≥99.0%

Supplier: TCI America

Synonyms: 2-Chlorobenzyl chloride1-Chloro-2-(chloromethyl)benzene

D0420-500G D0420-25G
TCD0420-500GEA 51.36 USD
TCD0420-500G TCD0420-25G
α,2-Dichlorotoluene ≥99.0%
α,2-Dichlorotoluene

CAS Number: 611-19-8
MDL Number: MFCD00000893
Molecular Formula: C7H6Cl2
Molecular Weight: 161.03
Purity/Analysis Method: 99.0% (GC)
Form: Clear Liquid
Boiling point (°C): 97
Flash Point (°C): 117
Specific Gravity (20/20): 1.28


Other Product Names / Synonyms: alpha,2-Dichlorotoluene

Formula: C₇H₆Cl₂
MW: 161.03 g/mol
Boiling Pt: 97 °C
Melting Pt: –13 °C
Density: 1.28 (20 °C)
Flash Pt: 117 °C
Storage Temperature: Ambient
MDL Number: MFCD00000893
CAS Number: 611-19-8
EINECS: 210-258-8
UN: 2235
ADR: 6.1,III
https://us.vwr.com/stibo/web/std.lang.all/49/49/28974949.jpg

Order Now

Specification Test Results

Purity/Analysis Method 99.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR