5,7-Dichloro-8-quinolinol ≥98.0% (by GC, titration analysis)

Synonyms: 5,7-Dichloro-8-hydroxyquinoline

D0412-25G D0412-5G
TCD0412-5GEA 65.92 USD
TCD0412-5G TCD0412-025G
5,7-Dichloro-8-quinolinol ≥98.0% (by GC, titration analysis)
5,7-Dichloro-8-quinolinol

CAS Number: 773-76-2
MDL Number: MFCD00006786
Molecular Formula: C9H5Cl2NO
Molecular Weight: 214.05
Purity/Analysis Method: 98.0% (GC,T)
Form: Crystal
Melting point (°C): 181


Product Categories: Materials Chemistry, Ligands for Functional Metal Complexes
Other Product Names / Synonyms: 5,7-Dichloro-8-oxine, 5,7-Dichloro-8-quinolinol

Formula: C₉H₅Cl₂NO
MW: 214.05 g/mol
Boiling Pt: 355 °C (1013 hPa)
Melting Pt: 181 °C
MDL Number: MFCD00006786
CAS Number: 773-76-2
EINECS: 212-258-3
Merck Index: 13,02193

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR