3,5-Dichloropyridine ≥98.0%

Supplier: TCI America

D0411-10G D0411-25G
TCD0411-025GEA 136.01 USD
TCD0411-025G TCD0411-010G
3,5-Dichloropyridine ≥98.0%
3,5-Dichloropyridine

CAS Number: 2457-47-8
MDL Number: MFCD00006376
Molecular Formula: C5H3Cl2N
Molecular Weight: 147.99
Purity/Analysis Method: 98.0% (GC)
Form: Crystal
Boiling point (°C): 178
Melting point (°C): 65

Formula: C₅H₃Cl₂N
MW: 147.99 g/mol
Boiling Pt: 178 °C
Melting Pt: 65 °C
MDL Number: MFCD00006376
CAS Number: 2457-47-8
EINECS: 219-537-9
UN: 2811
ADR: 6.1,III
https://us.vwr.com/stibo/web/std.lang.all/49/39/28974939.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR