1,3-Dichloro-2-propanol ≥98.0%
Supplier: TCI America
|
Danger
|
D0402-25G
TCD0402-25GEA
16.24
USD
TCD0402-25G
1,3-Dichloro-2-propanol ≥98.0%
1,3-Dichloro-2-propanol
CAS Number: 96-23-1
MDL Number: MFCD00000951
Molecular Formula: C3H6Cl2O
Molecular Weight: 128.98
Purity/Analysis Method:
Form: Clear Liquid
Boiling point (°C): 174
Melting point (°C): -4
Flash Point (°C): 74
Specific Gravity (20/20): 1.37
Other Product Names / Synonyms: alpha,alpha'-Dichlorohydrin, Glycerol alpha,gamma-Dichlorohydrin
Formula:
ClH₂CCHOHCH₂Cl MW: 128.99 g/mol Boiling Pt: 174 °C Melting Pt: –4 °C Density: 1.37 (20 °C) Flash Pt: 74 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00000951 CAS Number: 96-23-1 EINECS: 202-491-9 UN: 2750 ADR: 6.1,II Merck Index: 13,03100 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...