1,2-Dichloropropane ≥98.0%

Supplier: TCI America
Danger

Synonyms: Propylene dichloride

D0398-500G D0398-25G
TCD0398-500GEA 112.65 USD
TCD0398-500G TCD0398-25G
1,2-Dichloropropane ≥98.0%
1,2-Dichloropropane

CAS Number: 78-87-5
MDL Number: MFCD00000868
Molecular Formula: C3H6Cl2
Molecular Weight: 112.98
Purity/Analysis Method: 98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 97
Flash Point (°C): 14
Specific Gravity (20/20): 1.16


Other Product Names / Synonyms: Propylene Chloride, Propylene Dichloride

Formula: ClH₂CCH₂CH₂Cl
MW: 112.99 g/mol
Boiling Pt: 97 °C
Melting Pt: –100 °C
Density: 1.16 (20 °C)
Flash Pt: 14 °C
Storage Temperature: Ambient
MDL Number: MFCD00000868
CAS Number: 78-87-5
EINECS: 201-152-2
UN: 1279
ADR: 3,II
Merck Index: 12,08039
https://us.vwr.com/stibo/web/std.lang.all/49/12/28974912.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR