2,6-Dichlorophenol ≥99.0%

Supplier: TCI America
Warning

D0394-500G D0394-25G
TCD0394-500GEA 634.38 USD
TCD0394-500G TCD0394-25G
2,6-Dichlorophenol ≥99.0%
2,6-Dichlorophenol

CAS Number: 87-65-0
MDL Number: MFCD00002176
Molecular Formula: C6H4Cl2O
Molecular Weight: 163.00
Purity/Analysis Method: 99.0% (GC)
Form: Crystal
Boiling point (°C): 80
Melting point (°C): 66

Formula: C₆H₄Cl₂O
MW: 163 g/mol
Boiling Pt: 80 °C
Melting Pt: 66 °C
Density: 1.653 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00002176
CAS Number: 87-65-0
EINECS: 201-761-3
UN: 2020
ADR: 6.1,III
Merck Index: 13,03099
https://us.vwr.com/stibo/web/std.lang.all/49/04/28974904.jpg

Order Now

Specification Test Results

Purity/Analysis Method 99.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR