1,2-Dichloro-4-nitrobenzene ≥97.0%

Supplier: TCI America

D0388-500G D0388-25G
TCD0388-500GEA 94.74 USD
TCD0388-500G TCD0388-025G
1,2-Dichloro-4-nitrobenzene ≥97.0%
1,2-Dichloro-4-nitrobenzene

CAS Number: 99-54-7
MDL Number: MFCD00007207
Molecular Formula: C6H3Cl2NO2
Molecular Weight: 192.00
Purity/Analysis Method: 97.0% (GC)
Form: Crystal
Boiling point (°C): 255
Melting point (°C): 41
Flash Point (°C): 124


Other Product Names / Synonyms: 1,2-Dichloro-4-nitrobenzene

Formula: C₆H₃Cl₂NO₂
MW: 192 g/mol
Boiling Pt: 255 °C
Melting Pt: 41 °C
Density: 1.4558 g/cm³ (20 °C)
Flash Pt: 124 °C
Storage Temperature: Ambient
MDL Number: MFCD00007207
CAS Number: 99-54-7
EINECS: 202-764-2
UN: 1578
ADR: 6.1,II
https://us.vwr.com/stibo/web/std.lang.all/48/91/28974891.jpg

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR