TCIAD0358

D0358-25ML D0358-500ML D0358-100ML
TCD0358-25MLEA 23.3 USD
TCD0358-25ML TCD0358-500ML TCD0358-100ML
TCIAD0358
CAS Number: 75-78-5
MDL Number: MFCD00000491
Molecular Formula: C2H6Cl2Si
Molecular Weight: 129.06
Purity/Analysis Method: 98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 70
Melting point (°C): -76
Flash Point (°C): -7
Specific Gravity (20/20): 1.08
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR