2,4-Dichloroanisole ≥97.0%

Supplier: TCI America

Synonyms: 2,4-Dichloro-1-methoxybenzene

D0326-25G
TCD0326-025GEA 108.06 USD
TCD0326-025G
2,4-Dichloroanisole ≥97.0%
2,4-Dichloroanisole

CAS Number: 553-82-2
MDL Number: MFCD00044772
Molecular Formula: C7H6Cl2O
Molecular Weight: 177.02
Purity/Analysis Method: 97.0% (GC)
Form: Crystal
Freezing point (°C): 24


Other Product Names / Synonyms: 2,4-Dichloromethoxybenzene

Formula: C₇H₆Cl₂O
MW: 177.03 g/mol
Boiling Pt: 57 °C (0.2 mmHg)
Melting Pt: 28 °C
Density: 1.26 g/cm³ (20 °C)
MDL Number: MFCD00044772
CAS Number: 553-82-2
EINECS: 209-051-5
https://us.vwr.com/stibo/web/std.lang.all/47/97/28974797.jpg

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR