Dichloroacetyl chloride ≥98.0% (by GC, titration analysis)
Supplier: TCI America
|
Danger
|
D0313-500G
D0313-25G
TCD0313-500GEA
92.33
USD
TCD0313-500G
TCD0313-25G
Dichloroacetyl chloride ≥98.0% (by GC, titration analysis)
Dichloroacetyl chloride
CAS Number: 79-36-7
MDL Number: MFCD00000840
Molecular Formula: C2HCl3O
Molecular Weight: 147.38
Purity/Analysis Method:
Form: Clear Liquid
Boiling point (°C): 108
Flash Point (°C): 66
Specific Gravity (20/20): 1.53
Formula:
C₂HCl₃O MW: 147.39 g/mol Boiling Pt: 108 °C Density: 1.53 (20 °C) Flash Pt: 66 °C |
MDL Number:
MFCD00000840 CAS Number: 79-36-7 EINECS: 201-199-9 UN: 1765 ADR: 8,II Merck Index: 13,03078 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...