1,1-Diphenylethanol ≥98.0%
Supplier: TCI America
Synonyms:
α-Methylbenzhydryl alcohol
D0295-5G
D0295-25G
TCD0295-025GEA
150.22
USD
TCD0295-025G
TCD0295-005G
1,1-Diphenylethanol ≥98.0%
1,1-Diphenylethanol
CAS Number: 599-67-7
MDL Number: MFCD00004450
Molecular Formula: C14H14O
Molecular Weight: 198.27
Purity/Analysis Method:
Form: Crystal
Boiling point (°C): 155
Melting point (°C): 82
Formula:
(C₆H₅)₂C(OH)CH₃ MW: 198.26 g/mol Boiling Pt: 155 °C Melting Pt: 82 °C Density: 1.1059 g/cm³ (15 °C) |
MDL Number:
MFCD00004450 CAS Number: 599-67-7 EINECS: 209-970-1 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...