2-Nitro-m-xylene ≥99.0%

Supplier: TCI America

Synonyms: 1,3-Dimethyl-2-nitrobenzene

D0273-25G D0273-500G
TCD0273-25GEA 35.35 USD
TCD0273-25G TCD0273-500G
2-Nitro-m-xylene ≥99.0%
2-Nitro-m-xylene

CAS Number: 81-20-9
MDL Number: MFCD00007181
Molecular Formula: C8H9NO2
Molecular Weight: 151.17
Purity/Analysis Method: 99.0% (GC)
Form: Clear Liquid
Boiling point (°C): 222
Flash Point (°C): 87
Specific Gravity (20/20): 1.11


Other Product Names / Synonyms: 2-Nitro-m-xylene

Formula: C₈H₉NO₂
MW: 151.16 g/mol
Boiling Pt: 222 °C
Melting Pt: 14…16 °C
Density: 1.11 (20 °C)
Flash Pt: 87 °C
Storage Temperature: Ambient
MDL Number: MFCD00007181
CAS Number: 81-20-9
EINECS: 201-333-6
UN: 1665
ADR: 6.1,II
https://us.vwr.com/stibo/web/std.lang.all/47/22/28974722.jpg

Order Now

Specification Test Results

Purity/Analysis Method 99.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR