2,5-Di-tert-butyl-p-benzoquinone ≥98.0%
Supplier: TCI America
Synonyms:
2,5-Di-tert-butylcyclohexa-2,5-diene-1,4-dione
D0178-25G
TCD0178-025GEA
112.67
USD
TCD0178-025G
2,5-Di-tert-butyl-p-benzoquinone ≥98.0%
2,5-Di-tert-butyl-p-benzoquinone
CAS Number: 2460-77-7
MDL Number: MFCD00019442
Molecular Formula: C14H20O2
Molecular Weight: 220.31
Purity/Analysis Method:
Form: Crystal
Melting point (°C): 153
Other Product Names / Synonyms: 2,5-Di-tert-butyl-p-quinone
Formula:
C₁₄H₂₀O₂ MW: 220.31 g/mol Melting Pt: 153 °C |
MDL Number:
MFCD00019442 CAS Number: 2460-77-7 EINECS: 219-552-0 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...