1,10-Decanedithiol ≥98.0% (by GC, titration analysis)
Supplier: TCI America
|
Warning
|
Synonyms:
1,10-Dimercaptodecane
D0015-25G
TCD0015-025GEA
244.38
USD
TCD0015-025G
1,10-Decanedithiol ≥98.0% (by GC, titration analysis)
1,10-Decanedithiol
CAS Number: 1191-67-9
MDL Number: MFCD00022095
Molecular Formula: C10H22S2
Molecular Weight: 206.41
Purity/Analysis Method:
Form: Clear Liquid
Boiling point (°C): 297
Freezing point (°C): 17
Specific Gravity (20/20): 0.95
Product Categories: Materials Chemistry, Monomers
Other Product Names / Synonyms: 1,10-Dimercaptodecane
Formula:
C₁₀H₂₂S₂ MW: 206.42 g/mol Boiling Pt: 297 °C Melting Pt: 20 °C Density: 0.95 (20 °C) |
MDL Number:
MFCD00022095 CAS Number: 1191-67-9 EINECS: 214-739-3 UN: 3334 ADR: 9, |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...