Decahydronaphthalene (mixture of cis and trans isomers) ≥97.0%
Supplier: TCI America
|
Danger
|
Synonyms:
Bicyclo[4.4.0]decane
D0008-500ML
D0008-25ML
TCD0008-500MLEA
36.54
USD
TCD0008-500ML
TCD0008-25ML
Decahydronaphthalene (mixture of cis and trans isomers) ≥97.0%
Decahydronaphthalene (mixture of cis and trans isomers)
CAS Number: 91-17-8
MDL Number: MFCD00004130
Molecular Formula: C10H18
Molecular Weight: 138.25
Purity/Analysis Method:
Form: Clear Liquid
Boiling point (°C): 190
Flash Point (°C): 58
Specific Gravity (20/20): 0.89
Formula:
C₁₀H₁₈ MW: 138.25 g/mol Boiling Pt: 190 °C Melting Pt: ∼–40 °C Density: 0.89 (20 °C) Flash Pt: 58 °C Storage Temperature: Ambient |
MDL Number:
MFCD00004130 CAS Number: 91-17-8 EINECS: 202-046-9 UN: 1147 ADR: 3,III Merck Index: 13,02866 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...