Chloramphenicol ≥98.0% (by HPLC, titration analysis)
Supplier: TCI America
|
Danger
|
Synonyms:
Chloromycetin®
, (R,R)-Chloramphenicol
, Tega-Cetin
C2255-250G
C2255-25G
TCC2255-250GEA
293.21
USD
TCC2255-250G
TCC2255-25G
Chloramphenicol ≥98.0% (by HPLC, titration analysis)
Chloramphenicol
CAS Number: 56-75-7
MDL Number: MFCD00078159
Molecular Formula: C11H12Cl2N2O5
Molecular Weight: 323.13
Purity/Analysis Method:
Form: Crystal
Melting point (°C): 151
Specific rotation [a]20/D: 20 deg (C=5, EtOH)
Product Categories: Bioscience, Antibiotic Ingredients
Other Product Names / Synonyms: D-(-)-threo-2-Dichloroacetamido-1-(4-nitrophenyl)-1,3-propanediol, 2,2-Dichloro-N-[(1R,2R)-1,3-dihydroxy-1-(4-nitrophenyl)-2-propyl]acetamide
Formula:
Cl₂CHCONHCH(CH₂OH)CH(OH)C₆H₄NO₂ MW: 323.13 g/mol Boiling Pt: 645 °C (1013 hPa) Melting Pt: 151 °C Density: 1.49 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00078159 CAS Number: 56-75-7 EINECS: 200-287-4 Merck Index: 13,02087 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...