2-Chloro-4-fluorophenol ≥98.0%

Supplier: TCI America
Warning

C1598-25G
TCC1598-025GEA 179.21 USD
TCC1598-025G
2-Chloro-4-fluorophenol ≥98.0%
2-Chloro-4-fluorophenol

CAS Number: 1996-41-4
MDL Number: MFCD00002168
Molecular Formula: C6H4ClFO
Molecular Weight: 146.55
Purity/Analysis Method: 98.0% (GC)
Form: Crystal
Boiling point (°C): 172
Flash Point (°C): 75
Freezing point (°C): 21

Formula: C₆H₄ClFO
MW: 146.55 g/mol
Boiling Pt: 172 °C
Melting Pt: 23 °C
Density: 1.345 g/cm³ (20 °C)
Flash Pt: 75 °C
Storage Temperature: Ambient
MDL Number: MFCD00002168
CAS Number: 1996-41-4
EINECS: 217-876-7
UN: 3265
ADR: 8,III
https://us.vwr.com/stibo/web/std.lang.all/88/18/28938818.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR