1-Cyclohexyl-1-butanol ≥98.0%

Supplier: TCI America

Synonyms: α-Propylcyclohexanemethanol

C1568-5G
TCC1568-005GEA 37.54 USD
TCC1568-005G
1-Cyclohexyl-1-butanol ≥98.0%
1-Cyclohexyl-1-butanol

CAS Number: 4352-42-5
MDL Number: MFCD00019360
Molecular Formula: C10H20O
Molecular Weight: 156.27
Purity/Analysis Method: 98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 82
Flash Point (°C): 108
Specific Gravity (20/20): 0.91


Other Product Names / Synonyms: (1-Hydroxybutyl)cyclohexane

Formula: C₁₀H₂₀O
MW: 156.27 g/mol
Boiling Pt: 82 °C
Density: 0.91 (20 °C)
Flash Pt: 108 °C
MDL Number: MFCD00019360
CAS Number: 4352-42-5
EINECS: 224-419-5
https://us.vwr.com/stibo/web/std.lang.all/87/89/28938789.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR