N-(2-Chloroethyl)acetamide ≥98.0% (by titrimetric analysis)

Supplier: TCI America

C1489-5G
TCC1489-005GEA 147.95 USD
TCC1489-005G
N-(2-Chloroethyl)acetamide ≥98.0% (by titrimetric analysis)
N-(2-Chloroethyl)acetamide

CAS Number: 7355-58-0
MDL Number: MFCD00000965
Molecular Formula: C4H8ClNO
Molecular Weight: 121.56
Purity/Analysis Method: 98.0% (T)
Form: Clear Liquid
Boiling point (°C): 136
Flash Point (°C): 110
Specific Gravity (20/20): 1.18

Formula: C₄H₈ClNO
Boiling Pt: 136 °C
Density: 1.18 (20 °C)
Flash Pt: 110 °C
MDL Number: MFCD00000965
CAS Number: 7355-58-0
EINECS: 230-884-5
https://us.vwr.com/stibo/web/std.lang.all/87/08/28938708.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR