1-Chloro-3-methoxy-2-propanol ≥98.0%
Supplier: TCI America
Synonyms:
3-Chloro-1-methoxy-2-propanol
C1472-25G
TCC1472-025GEA
104.51
USD
TCC1472-025G
1-Chloro-3-methoxy-2-propanol ≥98.0%
1-Chloro-3-methoxy-2-propanol
CAS Number: 4151-97-7
MDL Number: MFCD00018935
Molecular Formula: C4H9ClO2
Molecular Weight: 124.56
Purity/Analysis Method:
Form: Clear Liquid
Boiling point (°C): 95
Flash Point (°C): 72
Specific Gravity (20/20): 1.17
Other Product Names / Synonyms: 3-Chloro-2-hydroxypropyl Methyl Ether
Formula:
C₄H₉ClO₂ Boiling Pt: 95 °C Density: 1.17 (20 °C) Flash Pt: 72 °C |
MDL Number:
MFCD00018935 CAS Number: 4151-97-7 EINECS: 223-982-4 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...