TCIAC1366

C1366-5G
TCC1366-005GEA 100.19 USD
TCC1366-005G
TCIAC1366
CAS Number: 601-57-0
MDL Number: MFCD00003663
Molecular Formula: C27H44O
Molecular Weight: 384.65
Purity/Analysis Method: 95.0% (GC)
Form: Crystal
Melting point (°C): 81
Specific rotation [a]20/D: 93 deg (C=2, CHCl3)

Product Categories: Bioscience, Steroids, Cholestanes
Other Product Names / Synonyms: (+)-3-Oxo-4-cholestene
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR