4-Chloro-4'-fluorobutyrophenone ≥97.0%

Supplier: TCI America

Synonyms: γ-Chloro-4-fluorobutyrophenoneω-Chloro-4-fluorobutyrophenone4-Chloro-1-(4-fluorophenyl)butan-1-one

C1294-25G
TCC1294-025GEA 44.63 USD
TCC1294-025G
4-Chloro-4'-fluorobutyrophenone ≥97.0%
4-Chloro-4'-fluorobutyrophenone

CAS Number: 3874-54-2
MDL Number: MFCD00001007
Molecular Formula: C10H10ClFO
Molecular Weight: 200.64
Purity/Analysis Method: 97.0% (GC)
Form: Clear Liquid
Boiling point (°C): 122
Flash Point (°C): 113
Specific Gravity (20/20): 1.23

Formula: C₁₀H₁₀ClFO
MW: 200.64 g/mol
Boiling Pt: 122 °C
Density: 1.23 (20 °C)
Flash Pt: 113 °C
MDL Number: MFCD00001007
CAS Number: 3874-54-2
EINECS: 223-395-3
https://us.vwr.com/stibo/web/std.lang.all/85/38/28938538.jpg

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR