D-Cycloserine ≥98.0% (by HPLC, titration analysis)
Supplier: TCI America
|
Warning
|
Synonyms:
(R)-(+)-Cycloserine
C1189-1G
C1189-5G
TCC1189-005GEA
203.93
USD
TCC1189-005G
TCC1189-001G
D-Cycloserine ≥98.0% (by HPLC, titration analysis)
D-Cycloserine
CAS Number: 68-41-7
MDL Number: MFCD00005353
Molecular Formula: C3H6N2O2
Molecular Weight: 102.09
Purity/Analysis Method:
Form: Crystal
Melting point (°C): 137
Specific rotation [a]20/D: 120 deg (C=1, H2O)
Storage Temperature: 0-10°C
Product Categories: Synthetic Organic Chemistry, Chiral Building Blocks
Other Product Names / Synonyms: (R)-(+)-4-Amino-3-isoxazolidinone, Orientomycin, Oxamycin
Formula:
C₃H₆N₂O₂ MW: 102.09 g/mol Melting Pt: 137 °C Storage Temperature: Freezer |
MDL Number:
MFCD00005353 CAS Number: 68-41-7 EINECS: 200-688-4 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...