3-Chlorobenzonitrile ≥98.0%

Supplier: TCI America
Warning

Synonyms: m-Chlorobenzonitrile

C1100-25G
TCC1100-025GEA 57.45 USD
TCC1100-025G
3-Chlorobenzonitrile ≥98.0%
3-Chlorobenzonitrile

CAS Number: 766-84-7
MDL Number: MFCD00001798
Molecular Formula: C7H4ClN
Molecular Weight: 137.57
Purity/Analysis Method: 98.0% (GC)
Form: Crystal
Boiling point (°C): 94
Melting point (°C): 43
Flash Point (°C): 97

Formula: C₇H₄ClN
MW: 137.57 g/mol
Boiling Pt: 94 °C
Melting Pt: 43 °C
Density: 1.14 g/cm³ (20 °C)
Flash Pt: 97 °C
Storage Temperature: Ambient
MDL Number: MFCD00001798
CAS Number: 766-84-7
EINECS: 212-172-6
UN: 3439
ADR: 6.1,III
https://us.vwr.com/stibo/web/std.lang.all/83/61/28938361.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR