Methyl carbazate ≥98.0% (by GC, titration analysis)

Supplier: TCI America

Synonyms: Methyl hydrazinocarboxylateMethyl hydrazinecarboxylate

C1098-500G C1098-25G
TCC1098-500GEA 176.61 USD
TCC1098-500G TCC1098-025G
Methyl carbazate ≥98.0% (by GC, titration analysis)
Methyl carbazate

CAS Number: 6294-89-9
MDL Number: MFCD00007594
Molecular Formula: C2H6N2O2
Molecular Weight: 90.08
Purity/Analysis Method: 98.0% (GC,T)
Form: Crystal
Boiling point (°C): 108
Melting point (°C): 73


Other Product Names / Synonyms: Carbazic Acid Methyl Ester, Methoxycarbonylhydrazine

Formula: C₂H₆N₂O₂
MW: 90.08 g/mol
Boiling Pt: 108 °C
Melting Pt: 73 °C
MDL Number: MFCD00007594
CAS Number: 6294-89-9
EINECS: 228-560-3
UN: 2811
ADR: 6.1,III
https://us.vwr.com/stibo/web/std.lang.all/83/59/28938359.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR