6-Chloro-2-hydroxypyridine ≥98.0% (by GC, titration analysis)

Supplier: TCI America
Warning

Synonyms: 6-Chloropyridin-2(1H)-one6-Chloro-2-pyridinol2-Chloro-6-pyridinol2-Chloro-6-hydroxypyridine

C1079-25G
TCC1079-025GEA 84.11 USD
TCC1079-025G
6-Chloro-2-hydroxypyridine ≥98.0% (by GC, titration analysis)
6-Chloro-2-hydroxypyridine

CAS Number: 16879-02-0
MDL Number: MFCD00006278
Molecular Formula: C5H4ClNO
Molecular Weight: 129.54
Purity/Analysis Method: 98.0% (GC,T)
Form: Crystal
Melting point (°C): 130
Flash Point (°C): 215


Other Product Names / Synonyms: 6-Chloro-2-pyridinol

Formula: C₅H₄ClNO
MW: 129.55 g/mol
Melting Pt: 130 °C
Flash Pt: 215 °C
MDL Number: MFCD00006278
CAS Number: 16879-02-0
EINECS: 240-909-1
https://us.vwr.com/stibo/web/std.lang.all/83/42/28938342.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR